1-Methoxy-2,2,2-trifluoroethanol, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: Trifluoroacetaldehyde methyl hemiacetal
PC7140-100G PC7140-25G
APOSPC7140-100GEA 1825 SEK
APOSPC7140-100G APOSPC7140-25G
1-Methoxy-2,2,2-trifluoroethanol, Technical Grade
1-Methoxy-2,2,2-trifluoroethanol
A stable source of trifluoroacetaldehyde. Add the product dropwise to PPA @150-180C - distill off & react at once.
Formel: C₃H₅F₃O₂
MW: 130,07 g/mol
Kokpunkt: 96 °C
Densitet: 1,26 g/cm³ (25 °C)
Flampunkt: 42 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013572
CAS nummer: 431-46-9
EINECS: 207-072-4
UN: 3271
ADR: 3,III