4-Borono-3-fluorothioanisole 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Fluoro-4-methylthiophenylboronic acid, (2-Fluoro-4-(methylthio)phenyl)boronic acid, 2-Fluoro-4-(methylsulphanyl)benzeneboronic acid
PC7124-250MG PC7124-500MG
APOSPC7124-250MGEA 1300 SEK
APOSPC7124-250MG APOSPC7124-500MG
4-Borono-3-fluorothioanisole 98%
4-Borono-3-fluorothioanisole
Formel: C₇H₈BFO₂S
MW: 186,0116 g/mol
Smältpunkt: 122…123 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09027247
CAS nummer: 957060-84-3