1-Ethoxy-2,2,2-trifluoroethanol, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: Trifluoroacetaldehyde ethyl hemiacetal, Trifluoroacetaldehydeethylhemiacetal
PC7120-100G PC7120-25G
APOSPC7120-100GEA 0 SEK
APOSPC7120-100G APOSPC7120-25G
1-Ethoxy-2,2,2-trifluoroethanol, Technical Grade
1-Ethoxy-2,2,2-trifluoroethanol
Formel: C₄H₇F₃O₂
MW: 144,09 g/mol
Kokpunkt: 104…105 °C/745mmHg
Densitet: 1,221 g/cm³ (25 °C)
Flampunkt: 39 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000441
CAS nummer: 433-27-2
EINECS: 207-087-6
UN: 1989
ADR: 3,III