Triethylsilyl trifluoromethanesulphonate 99%
Leverantör: Apollo Scientific
Fara
Synonyms: Triethylsilyl trifluoromethanesulphonate
PC7114-100G PC7114-25G PC7114-5G
APOSPC7114-100GEA 720 SEK
APOSPC7114-100G APOSPC7114-25G APOSPC7114-5G
Triethylsilyl trifluoromethanesulphonate 99%
Triethylsilyl trifluoromethanesulphonate
Lewis acid. Powerful silylating reagent .
Formel: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Kokpunkt: 207 °C
Densitet: 1.169 g/cm³ (25 °C)
Flampunkt: 66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000407
CAS nummer: 79271-56-0
EINECS: 279-124-4
UN: 3265
ADR: 8 II