5-Fluoronicotinonitrile 97+%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
3-Cyano-5-fluoropyridine
PC7108-1G
PC7108-250MG
APOSPC7108-1GEA
925
SEK
APOSPC7108-1G
APOSPC7108-250MG
5-Fluoronicotinonitrile 97+%
5-Fluoronicotinonitrile
Formel:
C₆H₃FN₂ MW: 122,10 g/mol Smältpunkt: 50…54 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD09027295 CAS nummer: 696-42-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...