1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene 98+%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Bromo-1-fluoro-4-(trifluoromethoxy)benzene
PC7089-5G PC7089-25G
APOSPC7089-5GEA 615 SEK
APOSPC7089-5G APOSPC7089-25G
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene 98+%
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene
Formel: C₇H₃BrF₄O
MW: 259.00 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02181186
CAS nummer: 286932-57-8