2-Fluoropyridine-5-carbonyl chloride 95%

Leverantör: Apollo Scientific
Fara

Synonyms: 6-Fluoronicotinoyl chloride

PC7055-1G PC7055-500MG PC7055-250MG
APOSPC7055-1GEA 1275 SEK
APOSPC7055-1G APOSPC7055-500MG APOSPC7055-250MG
2-Fluoropyridine-5-carbonyl chloride 95%
2-Fluoropyridine-5-carbonyl chloride
Formel: C₆H₃ClFNO
MW: 159,55 g/mol
Smältpunkt: 275…278 °C
Storage Temperature: Kylskåp
MDL Number: MFCD07781233
CAS nummer: 65352-94-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR