1,1,1-Trichlorotrifluoroacetone 97%
Leverantör: Apollo Scientific
|
Fara
|
PC7039-25G
PC7039-5G
APOSPC7039-25GEA
14350
SEK
APOSPC7039-25G
APOSPC7039-5G
1,1,1-Trichlorotrifluoroacetone 97%
1,1,1-Trichlorotrifluoroacetone
Formel:
Cl₃CC(O)CF₃ MW: 215.39 g/mol Kokpunkt: 86…90 °C Densitet: 1.59 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00042128 CAS nummer: 758-42-9 UN: 2922 ADR: 8+6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...