N-[4-(Trifluoromethoxy)phenyl]urea
Leverantör: Apollo Scientific
|
Varning
|
PC6935-25G
PC6935-5G
APOSPC6935-25GEA
790
SEK
APOSPC6935-25G
APOSPC6935-5G
N-[4-(Trifluoromethoxy)phenyl]urea
N-[4-(Trifluoromethoxy)phenyl]urea
Formel:
C₈H₇F₃N₂O₂ MW: 220,15 g/mol Smältpunkt: 153…155 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD03788552 CAS nummer: 82971-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...