4,5-Difluoro-2-iodobenzoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 4,5-Difluoro-2-Iodobenzoic acid
PC6916-1G PC6916-25G PC6916-5G
APOSPC6916-1GEA 175 SEK
APOSPC6916-1G APOSPC6916-25G APOSPC6916-5G
4,5-Difluoro-2-iodobenzoic acid
4,5-Difluoro-2-iodobenzoic acid
Formel: C₇H₃F₂IO₂
MW: 284 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02684316
CAS nummer: 130137-05-2