2,2,3,3-Tetrafluoropropyl methacrylate 97%
Leverantör: Apollo Scientific
Fara
Synonyms: Methacrylic Acid 2,2,3,3-Tetrafluoropropyl Ester
PC6869-100G PC6869-10G PC6869-25G
APOSPC6869-100GEA 775 SEK
APOSPC6869-100G APOSPC6869-10G APOSPC6869-25G
2,2,3,3-Tetrafluoropropyl methacrylate 97%
2,2,3,3-Tetrafluoropropyl methacrylate
Formel: C₇H₈F₄O₂
MW: 200,13 g/mol
Kokpunkt: 124 °C
Densitet: 1,25 g/cm³ (25 °C)
Flampunkt: 51 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00042381
CAS nummer: 45102-52-1
EINECS: 256-189-7
UN: 3272
ADR: 3,III