1H,1H,2H,2H-Perfluorooctyl isobutyrate 95%
Leverantör: Apollo Scientific
Varning
PC6526-25G PC6526-5G
APOSPC6526-25GEA 5575 SEK
APOSPC6526-25G APOSPC6526-5G
1H,1H,2H,2H-Perfluorooctyl isobutyrate 95%
1H,1H,2H,2H-Perfluorooctyl isobutyrate
Formel: C₁₂H₁₁F₁₃O₂
MW: 434,2 g/mol
Kokpunkt: 92 °C/14mmHg
Storage Temperature: Omgivande
MDL Number: MFCD01862031
CAS nummer: 242812-05-1