Silver trifluoromethanesulphonate 98%
Leverantör: Apollo Scientific
Fara
Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Trifluoromethanesulphonic acid silver salt
PC6450-25G PC6450-5G PC6450-100G PC6450-500G
APOSPC6450-25GEA 550 SEK
APOSPC6450-25G APOSPC6450-5G APOSPC6450-100G APOSPC6450-500G
Silver trifluoromethanesulphonate 98%
Silver trifluoromethanesulphonate
Increases leaving group ability of halogens; can be used for conversion of alkyl halides to triflates & acyl halides to acyl triflates; soluble in ether and other organic solvents.
Formel: CAgF₃O₃S
MW: 256,94 g/mol
Smältpunkt: 356…357 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013226
CAS nummer: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III