Perfluorosebacamide 97%

Leverantör: Apollo Scientific

PC6211-1G PC6211-5G
APOSPC6211-1GEA 3425 SEK
APOSPC6211-1G APOSPC6211-5G
Perfluorosebacamide 97%
Perfluorosebacamide
Formel: C₁₀H₄F₁₆N₂O₂
Smältpunkt: 233 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042364
CAS nummer: 307-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR