3-Amino-4-fluorobenzenesulphonic acid
Leverantör: Apollo Scientific
PC6146-1G
PC6146-500MG
APOSPC6146-1GEA
3175
SEK
APOSPC6146-1G
APOSPC6146-500MG
3-Amino-4-fluorobenzenesulphonic acid
3-Amino-4-fluorobenzenesulphonic acid
Formel:
C₆H₆FNO₃S Storage Temperature: Omgivande |
MDL Number:
MFCD12025945 CAS nummer: 349-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...