4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Leverantör: Apollo Scientific
Varning
PC6055-1G PC6055-5G
APOSPC6055-1GEA 1625 SEK
APOSPC6055-1G APOSPC6055-5G
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
4-{[5-(Trifluoromethyl)pyridin-2-yl]oxy}benzoic acid
Formel: C₁₃H₈F₃NO₃
Storage Temperature: Omgivande
MDL Number: MFCD00955325
CAS nummer: 773108-67-1