Perfluoro-1-heptene 95%
Leverantör: Apollo Scientific
Varning
PC6040-1G PC6040-5G
APOSPC6040-1GEA 1150 SEK
APOSPC6040-1G APOSPC6040-5G
Perfluoro-1-heptene 95%
Perfluoro-1-heptene
Contains up to 5% perfluorohept-2-ene
Formel: C₇F₁₄
MW: 350,05 g/mol
Kokpunkt: 80…82 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00013577
CAS nummer: 355-63-5