Perfluorododecane 97%

Leverantör: Apollo Scientific
Varning

PC5992D-1G PC5992D-5G PC5992D-250MG
APOSPC5992D-1GEA 0 SEK
APOSPC5992D-1G APOSPC5992D-5G APOSPC5992D-250MG
Perfluorododecane 97%
Perfluorododecane
Formel: C₁₂F₂₆
Kokpunkt: 178 °C
Smältpunkt: 75…77 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042084
CAS nummer: 307-59-5
EINECS: 206-204-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR