Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Dimethyl (3,3-difluoro-2-oxoheptyl)phosphonate, (3,3-Difluoro-2-oxoheptyl)phosphonic acid Dimethyl ester
PC5961-1G PC5961-5G
APOSPC5961-1GEA 855 SEK
APOSPC5961-1G APOSPC5961-5G
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate 97%
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate
Formel: C₉H₁₇F₂O₄P
MW: 258,20 g/mol
Kokpunkt: 108…110 °C/0,4mmHg
Storage Temperature: Omgivande
MDL Number: MFCD11707108
CAS nummer: 50889-46-8