Perfluorocyclohexane 90%

Leverantör: Apollo Scientific
Varning

PC5930-100MG PC5930-1G PC5930-250MG
APOSPC5930-100MGEA 1625 SEK
APOSPC5930-100MG APOSPC5930-1G APOSPC5930-250MG
Perfluorocyclohexane 90%
Perfluorocyclohexane
Formel: C₆F₁₂
MW: 300,05 g/mol
Kokpunkt: 59…60 °C
Smältpunkt: 51 °C (subl.)
Densitet: 1,684 g/cm³ (25 °C)
Storage Temperature: Frys
MDL Number: MFCD00003821
CAS nummer: 355-68-0
EINECS: 206-591-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR