1H,1H,2H,2H-Nonafluorohexyl methacrylate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Methacrylic acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl methacrylate, Methacrylic acid 1H,1H,2H,2H-Nonafluorohexyl ester
PC5906E-1G PC5906E-25G PC5906E-5G
APOSPC5906E-1GEA 345 SEK
APOSPC5906E-1G APOSPC5906E-25G APOSPC5906E-5G
1H,1H,2H,2H-Nonafluorohexyl methacrylate 97%
1H,1H,2H,2H-Nonafluorohexyl methacrylate
Formel: C₁₀H₉F₉O₂
Kokpunkt: 60…62 °C/5mmHg
Densitet: 1,402 g/cm³ (25 °C)
Flampunkt: 77 °C
Storage Temperature: Omgivande
MDL Number: MFCD00236094
CAS nummer: 1799-84-4