3-Fluoro-5-(trifluoromethyl)cinnamic acid
Leverantör: Apollo Scientific
Varning
PC5867-1G PC5867-5G
APOSPC5867-1GEA 260 SEK
APOSPC5867-1G APOSPC5867-5G
3-Fluoro-5-(trifluoromethyl)cinnamic acid
3-Fluoro-5-(trifluoromethyl)cinnamic acid
Formel: C₁₀H₆F₄O₂
Storage Temperature: Omgivande
MDL Number: MFCD00236296
CAS nummer: 575469-96-4