3-Chloro-5-(trifluoromethoxy)benzyl bromide 97%
Leverantör: Apollo Scientific
Fara
PC5790-1G PC5790-5G
APOSPC5790-1GEA 1375 SEK
APOSPC5790-1G APOSPC5790-5G
3-Chloro-5-(trifluoromethoxy)benzyl bromide 97%
3-Chloro-5-(trifluoromethoxy)benzyl bromide
Formel: C₈H₅BrClF₃O
MW: 289.48 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06660311
CAS nummer: 886503-33-9
UN: 3265
ADR: 8 II