Ethyl-2,6-difluorobenzoate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Ethyl 2,6-difluorobenzoate
PC57044-100G PC57044-25G PC57044-5G
APOSPC57044-100GEA 2500 SEK
APOSPC57044-100G APOSPC57044-25G APOSPC57044-5G
Ethyl-2,6-difluorobenzoate 97%
Ethyl-2,6-difluorobenzoate
Formel: C₉H₈F₂O₂
MW: 186,16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06204338
CAS nummer: 19064-14-3