3-Amino-4-fluoro-1-methyl-1H-indazole
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
3-Amino-4-fluoro-1-methylindazole
PC5685-250MG
PC5685-100MG
APOSPC5685-250MGEA
855
SEK
APOSPC5685-250MG
APOSPC5685-100MG
3-Amino-4-fluoro-1-methyl-1H-indazole
3-Amino-4-fluoro-1-methyl-1H-indazole
Formel:
C₈H₈FN₃ MW: 165,1700 g/mol Smältpunkt: 125…127 °C Storage Temperature: Omgivande |
MDL Number:
MFCD11109397 CAS nummer: 162502-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...