3-(Trifluoromethyl)phenylboronic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (3-(Trifluoromethyl)phenyl)boronic acid, (3-(Trifluoromethyl)phenyl)metaboric acid, 3-(Trifluoromethyl)benzeneboronic acid, Dihydroxy(3-(trifluoromethyl)phenyl)borane
PC5641-100G PC5641-25G PC5641-5G
APOSPC5641-100GEA 1550 SEK
APOSPC5641-100G APOSPC5641-25G APOSPC5641-5G
3-(Trifluoromethyl)phenylboronic acid 98%
3-(Trifluoromethyl)phenylboronic acid
Formel: C₇H₆BF₃O₂
MW: 189,93 g/mol
Smältpunkt: 162…164 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151854
CAS nummer: 1423-26-3