5,6-Difluoro-2,1,3-benzothiadiazole

Leverantör: Apollo Scientific
Varning

Synonyms: 5,6-Difluorobenzo[c][1,2,5]thiadiazole

PC56068-1G PC56068-250MG
APOSPC56068-1GEA 1175 SEK
APOSPC56068-1G APOSPC56068-250MG
5,6-Difluoro-2,1,3-benzothiadiazole
5,6-Difluoro-2,1,3-benzothiadiazole
Formel: C₆H₂F₂N₂S
MW: 172,15 g/mol
Kokpunkt: 218,2 °C
Flampunkt: 87,5 °C
Storage Temperature: Omgivande
MDL Number: MFCD28100486
CAS nummer: 1293389-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR