1,1,1,2,2-Pentafluoro-3-oxopentane 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Pentafluoroethyl ethyl ketone
PC5570-5G PC5570-1G
APOSPC5570-5GEA 9950 SEK
APOSPC5570-5G APOSPC5570-1G
1,1,1,2,2-Pentafluoro-3-oxopentane 97%
1,1,1,2,2-Pentafluoro-3-oxopentane
Formel: C₅H₅F₅O
MW: 176,09 g/mol
Kokpunkt: 61…62 °C/740mmHg
Densitet: 1,554 g/cm³ (25 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00053172
CAS nummer: 378-72-3
UN: 1993
ADR: 3,III