2,6-Difluorophenyl isothiocyanate 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 2,6-Difluorophenyl isocyanate, 2,6-Difluorophenylisothiocyanate, 1,3-Difluoro-2-isothiocyanatobenzene
PC5347-1G PC5347-5G
APOSPC5347-1GEA 175 SEK
APOSPC5347-1G APOSPC5347-5G
2,6-Difluorophenyl isothiocyanate 97%
2,6-Difluorophenyl isothiocyanate
Formel: C₇H₃F₂NS
MW: 171,17 g/mol
Kokpunkt: 217 °C
Densitet: 1,325 g/cm³ (25 °C)
Flampunkt: 104 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00041047
CAS nummer: 207974-17-2
UN: 1760
ADR: 8,II