4-Chloro-3-iodobenzotrifluoride, Technical Grade
Leverantör: Apollo Scientific
Fara
Synonyms: 4-Chloro-3-iodobenzotrifluoride, 1-Chloro-2-iodo-4-(trifluoromethyl)benzene, 4-Chloro-α-α-α-trifluoro-3-iodotoluene
PC53433-100G PC53433-1KG PC53433-500G
APOSPC53433-100GEA 550 SEK
APOSPC53433-100G APOSPC53433-1KG APOSPC53433-500G
4-Chloro-3-iodobenzotrifluoride, Technical Grade
4-Chloro-3-iodobenzotrifluoride
Formel: C₇H₃ClF₃I
MW: 306.45 g/mol
Kokpunkt: 58
Densitet: 1.92
Storage Temperature: Kylskåp
MDL Number: MFCD00001037
CAS nummer: 672-57-1
UN: 3265
ADR: 8 II