2-Bromo-4,5-diluorotoluene
Leverantör: Apollo Scientific
Varning
Synonyms: 4,5-Difluoro-2-methylbromobenzene, 1-Bromo-3,4-difluoro-6-methylbenzene, 1-Bromo-4,5-difluoro-2-methylbenzene
PC53115-1G PC53115-5G
APOSPC53115-1GEA 825 SEK
APOSPC53115-1G APOSPC53115-5G
2-Bromo-4,5-diluorotoluene
2-Bromo-4,5-diluorotoluene
Formel: C₇H₅BrF₂
MW: 207,02 g/mol
Densitet: 1,62 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD07777160
CAS nummer: 875664-38-3