Nitronium tetrafluoroborate 96%
Leverantör: Apollo Scientific
Fara
Synonyms: Nitryl tetrafluoroborate
PC5270-100G PC5270-25G PC5270-5G
APOSPC5270-100GEA 8775 SEK
APOSPC5270-100G APOSPC5270-25G APOSPC5270-5G
Nitronium tetrafluoroborate 96%
Nitronium tetrafluoroborate
Powerful nitrating agent
Formel: BF₄NO₂
MW: 132,81 g/mol
Smältpunkt: 240 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00011432
CAS nummer: 13826-86-3
EINECS: 237-533-5
UN: 3260
ADR: 8,II