Diethyl (difluoromethyl)phosphonate
Leverantör: Apollo Scientific
Varning
Synonyms: Difluoromethylphosphonicaciddiethylester
PC52115-1G PC52115-500MG
APOSPC52115-1GEA 260 SEK
APOSPC52115-1G APOSPC52115-500MG
Diethyl (difluoromethyl)phosphonate
Diethyl (difluoromethyl)phosphonate
Formel: C₅H₁₁F₂O₃P
Kokpunkt: 55 °C
Densitet: 1,2059 g/cm³ (25 °C)
Flampunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00192023
CAS nummer: 1478-53-1