3'-Bromo-5'-(trifluoromethyl)acetophenone 99%
Leverantör: Apollo Scientific
Synonyms: Ethanone, 1-[3-bromo-5-(trifluoromethyl)phenyl]-, 1-[3-Bromo-5-(trifluoromethyl)phenyl]ethanone
PC520654-1G PC520654-5G
APOSPC520654-1GEA 825 SEK
APOSPC520654-1G APOSPC520654-5G
3'-Bromo-5'-(trifluoromethyl)acetophenone 99%
3'-Bromo-5'-(trifluoromethyl)acetophenone
Formel: C₉H₆BrF₃O
MW: 267.04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11847339
CAS nummer: 154259-25-3