5-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%

Leverantör: Apollo Scientific

PC520621-10G PC520621-1G PC520621-5G
APOSPC520621-10GEA 13550 SEK
APOSPC520621-10G APOSPC520621-1G APOSPC520621-5G
5-(Trifluoromethyl)pyridine-2-carbonyl chloride 97%
5-(Trifluoromethyl)pyridine-2-carbonyl chloride
Formel: C₇H₃ClF₃NO
Storage Temperature: Omgivande
MDL Number: MFCD11226644
CAS nummer: 128073-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR