Methyl-2-chloro-4-fluorobenzoate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 2-chloro-4-fluorobenzenecarboxylate, Methyl 2-chloro-4-fluorobenzoate, 2-Chloro-4-fluorobenzoic acid methyl ester
PC5074-100G PC5074-25G PC5074-5G
APOSPC5074-100GEA 1125 SEK
APOSPC5074-100G APOSPC5074-25G APOSPC5074-5G
Methyl-2-chloro-4-fluorobenzoate 97%
Methyl-2-chloro-4-fluorobenzoate
Formel: C₈H₆ClFO₂
MW: 188,59 g/mol
Kokpunkt: 103…105 °C/15mmHg
Storage Temperature: Omgivande
MDL Number: MFCD00173937
CAS nummer: 85953-29-3