α,α,α-Trifluoro-o-xylene 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Methylbenzotrifluoride, 1-Methyl-2-(trifluoromethyl)benzene, 2-(Trifluoromethyl)toulene
PC5067-1G PC5067-25G PC5067-5G
APOSPC5067-1GEA 175 SEK
APOSPC5067-1G APOSPC5067-25G APOSPC5067-5G
α,α,α-Trifluoro-o-xylene 97%
α,α,α-Trifluoro-o-xylene
Formel: C₈H₇F₃
MW: 160,14 g/mol
Kokpunkt: 125…126 °C
Densitet: 1,174 g/cm³ (25 °C)
Flampunkt: 26 °C
Storage Temperature: Omgivande
MDL Number: MFCD00153204
CAS nummer: 13630-19-8
UN: 1993
ADR: 3,III