(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
Leverantör: Apollo Scientific
Fara
Synonyms: (+)-Mosher's Acid Chloride, (+)-MTPA-Cl, (S)-(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride, (S)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoyl chloride
PC5063J-1G PC5063J-250MG
APOSPC5063J-1GEA 2275 SEK
APOSPC5063J-1G APOSPC5063J-250MG
(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 99%
(+)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride
Formel: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Kokpunkt: 80 °C/14mmHg
Densitet: 1.34 g/cm³ (25 °C)
Flampunkt: 89 °C
Storage Temperature: Frys
MDL Number: MFCD00067105
CAS nummer: 20445-33-4
UN: 3265
ADR: 8 III