3-Ethoxy-5-(trifluoromethyl)phenylboronic acid 98%
Leverantör: Apollo Scientific
Synonyms: 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Ethoxy-5-trifluoromethylphenylboronic acid
PC5051-1G PC5051-5G
APOSPC5051-1GEA 2150 SEK
APOSPC5051-1G APOSPC5051-5G
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid 98%
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid
Formel: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
Smältpunkt: 173…176 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08056361
CAS nummer: 871332-96-6