Methyl-3-bromo-5-fluorobenzoate
Leverantör: Apollo Scientific
Synonyms: Methyl 3-bromo-5-fluorobenzoate
PC501800-25G PC501800-5G PC501800-100G
APOSPC501800-25GEA 2350 SEK
APOSPC501800-25G APOSPC501800-5G APOSPC501800-100G
Methyl-3-bromo-5-fluorobenzoate
Methyl-3-bromo-5-fluorobenzoate
Formel: C₈H₆BrFO₂
MW: 233,04 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD07780735
CAS nummer: 334792-52-8