1-Bromo-3-(2,2-difluoroethoxy)benzene
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(2,2-Difluoroethoxy)bromobenzene
PC500915-1G PC500915-250MG PC500915-100MG
APOSPC500915-1GEA 7325 SEK
APOSPC500915-1G APOSPC500915-250MG APOSPC500915-100MG
1-Bromo-3-(2,2-difluoroethoxy)benzene
1-Bromo-3-(2,2-difluoroethoxy)benzene
Formel: C₈H₇BrF₂O
Storage Temperature: Omgivande
MDL Number: MFCD11036117
CAS nummer: 958454-25-6