Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 95%
Leverantör: Apollo Scientific
Fara
Synonyms: Ethyl 2,2-difluoro-2-(trimethylsilyl)acetate, 2,2-Difluoro-2-(trimethylsilyl)acetic acid Ethyl ester
PC49333-25G PC49333-1G PC49333-5G
APOSPC49333-25GEA 5150 SEK
APOSPC49333-25G APOSPC49333-1G APOSPC49333-5G
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate 95%
Ethyl 2,2-Difluoro-2-(trimethylsilyl)acetate
Formel: C₇H₁₄F₂O₂Si
MW: 196,27 g/mol
Kokpunkt: 55 °C/20mmHg
Storage Temperature: Omgivande
MDL Number: MFCD04973092
CAS nummer: 205865-67-4
UN: 1993
ADR: 3,III