2-Iodononafluorobutane 98%
Leverantör: Apollo Scientific
|
Varning
|
PC4928-25G
PC4928-5G
APOSPC4928-25GEA
0
SEK
APOSPC4928-25G
APOSPC4928-5G
2-Iodononafluorobutane 98%
2-Iodononafluorobutane
Formel:
C₄F₉I Kokpunkt: 66 °C Densitet: 2,095 g/cm³ (25 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00077556 CAS nummer: 375-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...