5-Chloro-2,4-difluorobenzoic acid 97%
Leverantör: Apollo Scientific
Varning
PC49159-1G PC49159-5G
APOSPC49159-1GEA 175 SEK
APOSPC49159-1G APOSPC49159-5G
5-Chloro-2,4-difluorobenzoic acid 97%
5-Chloro-2,4-difluorobenzoic acid
Formel: C7H3ClF2O2
MW: 192,5473 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06658163
CAS nummer: 130025-33-1