4-Methoxy-2-(trifluoromethoxy)benzaldehyde
Leverantör: Apollo Scientific
Varning
PC49158-10G PC49158-5G
APOSPC49158-10GEA 3600 SEK
APOSPC49158-10G APOSPC49158-5G
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
4-Methoxy-2-(trifluoromethoxy)benzaldehyde
Formel: C₉H₇F₃O₃
MW: 220.15 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06660317
CAS nummer: 886503-52-2