2-(Difluoromethyl)-1,4-difluorobenzene 97%
Leverantör: Apollo Scientific
Synonyms: 2,5-Difluorobenzal fluoride
PC49149-1G PC49149-25G PC49149-5G
APOSPC49149-1GEA 1400 SEK
APOSPC49149-1G APOSPC49149-25G APOSPC49149-5G
2-(Difluoromethyl)-1,4-difluorobenzene 97%
2-(Difluoromethyl)-1,4-difluorobenzene
Formel: C₇H₄F₄
MW: 164,1 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12026289
CAS nummer: 195886-79-4