3',5'-Dichloro-2,2,2-trifluoroacetophenone 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethanone
PC48738-1G PC48738-25G PC48738-5G
APOSPC48738-1GEA 175 SEK
APOSPC48738-1G APOSPC48738-25G APOSPC48738-5G
3',5'-Dichloro-2,2,2-trifluoroacetophenone 98%
3',5'-Dichloro-2,2,2-trifluoroacetophenone
Formel: C₈H₃Cl₂F₃O
MW: 243,0100 g/mol
Densitet: 1,51 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD01319996
CAS nummer: 130336-16-2