4-Chloro-2-fluoro-5-(methoxycarbonyl)phenylboronic acid pinacol ester
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 2-chloro-4-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
PC48622-1G PC48622-250MG
APOSPC48622-1GEA 7700 SEK
APOSPC48622-1G APOSPC48622-250MG
4-Chloro-2-fluoro-5-(methoxycarbonyl)phenylboronic acid pinacol ester
4-Chloro-2-fluoro-5-(methoxycarbonyl)phenylboronic acid pinacol ester
Formel: C₁₄H₁₇BClFO₄
MW: 314.5500 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD12026085
CAS nummer: 1073339-13-5