4-Fluorobenzoyl chloride, Technical Grade

Leverantör: Apollo Scientific
Fara

Synonyms: p-Fluorobenzoyl chloride

PC48430-100G PC48430-250G
APOSPC48430-100GEA 175 SEK
APOSPC48430-100G APOSPC48430-250G
4-Fluorobenzoyl chloride, Technical Grade
4-Fluorobenzoyl chloride
Formel: C₇H₄ClFO
MW: 158,56 g/mol
Kokpunkt: 82 °C/20mmHg
Smältpunkt: 44540 °C
Densitet: 1,326 g/cm³ (25 °C)
Flampunkt: 82 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000684
CAS nummer: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR