Dihydrogen hexafluorozirconate 45% (w/v) i vattenlösning
Leverantör: Apollo Scientific
Synonyms: Zirconate(2-), hexafluoro-, Hexafluorozirconic acid, Hexafluorozirconate(2-)
PC4812-2KG PC4812-500G
APOSPC4812-2KGEA 3400 SEK
APOSPC4812-2KG APOSPC4812-500G
Dihydrogen hexafluorozirconate 45% (w/v) i vattenlösning
Dihydrogen hexafluorozirconate
Formel: H₂ZrF₆
MW: 205,215 Da
Storage Temperature: Omgivande
MDL Number: MFCD00082965
CAS nummer: 12021-95-3